Anti-PRSS27 Rat Monoclonal Antibody [clone: 246407]

Supplier: R & D Systems
MAB1989
220019-506EA 772.25 USD
220019-506
Anti-PRSS27 Rat Monoclonal Antibody [clone: 246407]
Antibodies
Type: Primary
Antigen: PRSS27
Clonality: Monoclonal
Clone: 246407
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR