Anti-PRSS27 Mouse Monoclonal Antibody [clone: 243305]

Supplier: R & D Systems
MAB1988
220019-505EA 763.57 USD
220019-505
Anti-PRSS27 Mouse Monoclonal Antibody [clone: 243305]
Antibodies
Type: Primary
Antigen: PRSS27
Clonality: Monoclonal
Clone: 243305
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR