Anti-CER1 Rat Monoclonal Antibody [clone: 225807]

Supplier: R & D Systems
MAB1986
220019-504EA 763.57 USD
220019-504
Anti-CER1 Rat Monoclonal Antibody [clone: 225807]
Antibodies
Type: Primary
Antigen: CER1
Clonality: Monoclonal
Clone: 225807
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR