Anti-PRKRA Mouse Monoclonal Antibody [clone: HL71/10]

Supplier: R & D Systems
MAB1980
220019-499EA 655.11 USD
220019-499
Anti-PRKRA Mouse Monoclonal Antibody [clone: HL71/10]
Antibodies
Type: Primary
Antigen: PRKRA
Clonality: Monoclonal
Clone: HL71/10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR