Anti-FZD7 Rat Monoclonal Antibody [clone: 151107]

Supplier: R & D Systems
MAB198-500 MAB198-050
220019-503EA 815.63 USD
220019-503 220019-500
Anti-FZD7 Rat Monoclonal Antibody [clone: 151107]
Antibodies
Type: Primary
Antigen Symbol: FZD7
Antigen Name: frizzled homolog 7 (Drosophila)
Antigen Synonyms: Fz7
Clonality: Monoclonal
Clone: 151107
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR