Anti-OTX2 Mouse Monoclonal Antibody [clone: 246826]

Supplier: R & D Systems
MAB1979
220019-498EA 594.37 USD
220019-498
Anti-OTX2 Mouse Monoclonal Antibody [clone: 246826]
Antibodies
Type: Primary
Antigen: OTX2
Clonality: Monoclonal
Clone: 246826
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR