Anti-MIF Rat Monoclonal Antibody [clone: 811409]

Supplier: R & D Systems
MAB19781
102885-420EA 594.37 USD
102885-420
Anti-MIF Rat Monoclonal Antibody [clone: 811409]
Antibodies
Type: Primary
Antigen: MIF
Clonality: Monoclonal
Clone: 811409
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgM
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR