Anti-MIF Rat Monoclonal Antibody [clone: 811429]

Supplier: R & D Systems
MAB1978
102885-418EA 577.02 USD
102885-418
Anti-MIF Rat Monoclonal Antibody [clone: 811429]
Antibodies
Type: Primary
Antigen: MIF
Clonality: Monoclonal
Clone: 811429
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR