Anti-IL11RA Mouse Monoclonal Antibody [clone: 473143]

Supplier: R & D Systems
MAB1977
220019-497EA 655.11 USD
220019-497
Anti-IL11RA Mouse Monoclonal Antibody [clone: 473143]
Antibodies
Type: Primary
Antigen: IL11RA
Clonality: Monoclonal
Clone: 473143
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR