Anti-NOV Rat Monoclonal Antibody [clone: 231226]

Supplier: R & D Systems
MAB19762
220019-496EA 763.57 USD
220019-496
Anti-NOV Rat Monoclonal Antibody [clone: 231226]
Antibodies
Type: Primary
Antigen: NOV
Clonality: Monoclonal
Clone: 231226
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR