Anti-NOV Rat Monoclonal Antibody [clone: 231216]

Supplier: R & D Systems
MAB1976
220019-495EA 763.57 USD
220019-495
Anti-NOV Rat Monoclonal Antibody [clone: 231216]
Antibodies
Type: Primary
Antigen: NOV
Clonality: Monoclonal
Clone: 231216
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR