Anti-CCR7 Mouse Monoclonal Antibody [clone: 150503]

Supplier: R & D Systems
MAB197-100
102885-416EA 655.11 USD
102885-416
Anti-CCR7 Mouse Monoclonal Antibody [clone: 150503]
Antibodies
Type: Primary
Antigen: CCR7
Clonality: Monoclonal
Clone: 150503
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR