Anti-CD22 Mouse Monoclonal Antibody [clone: 219902]

Supplier: R & D Systems
MAB19681
102885-414EA 594.37 USD
102885-414
Anti-CD22 Mouse Monoclonal Antibody [clone: 219902]
Antibodies
Type: Primary
Antigen: CD22
Clonality: Monoclonal
Clone: 219902
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR