Anti-CD22 Mouse Monoclonal Antibody [clone: 219934]

Supplier: R & D Systems
MAB1968
220019-492EA 594.37 USD
220019-492
Anti-CD22 Mouse Monoclonal Antibody [clone: 219934]
Antibodies
Type: Primary
Antigen: CD22
Clonality: Monoclonal
Clone: 219934
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR