Anti-IL24 Mouse Monoclonal Antibody [clone: 283161]

Supplier: R & D Systems
MAB19652
220019-491EA 772.25 USD
220019-491
Anti-IL24 Mouse Monoclonal Antibody [clone: 283161]
Antibodies
Type: Primary
Antigen: IL24
Clonality: Monoclonal
Clone: 283161
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR