Anti-IL24 Mouse Monoclonal Antibody [clone: 244202]

Supplier: R & D Systems
MAB1965
220019-489EA 763.57 USD
220019-489
Anti-IL24 Mouse Monoclonal Antibody [clone: 244202]
Antibodies
Type: Primary
Antigen: IL24
Clonality: Monoclonal
Clone: 244202
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR