Anti-FCGR3 Rat Monoclonal Antibody [clone: 275024]

Supplier: R & D Systems
MAB1960
220019-487EA 577.02 USD
220019-487
Anti-FCGR3 Rat Monoclonal Antibody [clone: 275024]
Antibodies
Type: Primary
Antigen Symbol: FCGR3
Antigen Name: Fc receptor, IgG, low affinity III
Antigen Synonyms: CD16
Clonality: Monoclonal
Clone: 275024
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR