Anti-DHH Rat Monoclonal Antibody [clone: 148644]

Supplier: R & D Systems
MAB196
220019-486EA 763.57 USD
220019-486
Anti-DHH Rat Monoclonal Antibody [clone: 148644]
Antibodies
Type: Primary
Antigen: DHH
Clonality: Monoclonal
Clone: 148644
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR