Anti-IL12RB2 Mouse Monoclonal Antibody [clone: 622509]

Supplier: R & D Systems
MAB19591
102885-412EA 594.37 USD
102885-412
Anti-IL12RB2 Mouse Monoclonal Antibody [clone: 622509]
Antibodies
Type: Primary
Antigen: IL12RB2
Clonality: Monoclonal
Clone: 622509
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR