Anti-GDF11 Mouse Monoclonal Antibody [clone: 743835]

Supplier: R & D Systems
MAB19582
102885-410EA 600.15 USD
102885-410
Anti-GDF11 Mouse Monoclonal Antibody [clone: 743835]
Antibodies
Type: Primary
Antigen Symbol: GDF11
Antigen Name: growth differentiation factor 11
Antigen Synonyms: BMP-11, BMP11
Clonality: Monoclonal
Clone: 743835
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR