Anti-FGF7 Mouse Monoclonal Antibody [clone: 299026]

Supplier: R & D Systems
MAB1957
220019-484EA 561.11 USD
220019-484
Anti-FGF7 Mouse Monoclonal Antibody [clone: 299026]
Antibodies
Type: Primary
Antigen Symbol: FGF7
Antigen Name: fibroblast growth factor 7
Antigen Synonyms: KGF
Clonality: Monoclonal
Clone: 299026
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR