Anti-CD36 Mouse Monoclonal Antibody [clone: 877346]

Supplier: R & D Systems
MAB19552
102885-406EA 594.37 USD
102885-406
Anti-CD36 Mouse Monoclonal Antibody [clone: 877346]
Antibodies
Simple Western

Type: Primary
Antigen: CD36
Clonality: Monoclonal
Clone: 877346
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR