Anti-CD36 Rat Monoclonal Antibody [clone: 255606]

Supplier: R & D Systems
MAB19551
220019-482EA 779.48 USD
220019-482
Anti-CD36 Rat Monoclonal Antibody [clone: 255606]
Antibodies
Type: Primary
Antigen: CD36
Clonality: Monoclonal
Clone: 255606
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR