Anti-CD36 Rat Monoclonal Antibody [clone: 255619]

Supplier: R & D Systems
MAB1955
220019-481EA 577.02 USD
220019-481
Anti-CD36 Rat Monoclonal Antibody [clone: 255619]
Antibodies
Type: Primary
Antigen: CD36
Clonality: Monoclonal
Clone: 255619
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR