Anti-ICAM5 Mouse Monoclonal Antibody [clone: 283607]

Supplier: R & D Systems
MAB1950
220019-480EA 616.06 USD
220019-480
Anti-ICAM5 Mouse Monoclonal Antibody [clone: 283607]
Antibodies
Type: Primary
Antigen: ICAM5
Clonality: Monoclonal
Clone: 283607
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR