Anti-CCR6 Mouse Monoclonal Antibody [clone: 53103]

Supplier: R & D Systems
MAB195-100
102885-404EA 639.2 USD
102885-404
Anti-CCR6 Mouse Monoclonal Antibody [clone: 53103]
Antibodies
Type: Primary
Antigen: CCR6
Clonality: Monoclonal
Clone: 53103
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR