Anti-APCS Mouse Monoclonal Antibody [clone: 91119]

Supplier: R & D Systems
MAB1948
102885-402EA 594.37 USD
102885-402
Anti-APCS Mouse Monoclonal Antibody [clone: 91119]
Antibodies
Type: Primary
Antigen: APCS
Clonality: Monoclonal
Clone: 91119
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR