Anti-KREMEN2 Mouse Monoclonal Antibody [clone: 276709]

Supplier: R & D Systems
MAB1946
220019-478EA 577.02 USD
220019-478
Anti-KREMEN2 Mouse Monoclonal Antibody [clone: 276709]
Antibodies
Type: Primary
Antigen: KREMEN2
Clonality: Monoclonal
Clone: 276709
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR