Anti-IL13 Mouse Monoclonal Antibody [clone: 296202]

Supplier: R & D Systems
MAB1945
220019-477EA 577.02 USD
220019-477
Anti-IL13 Mouse Monoclonal Antibody [clone: 296202]
Antibodies
Type: Primary
Antigen: IL13
Clonality: Monoclonal
Clone: 296202
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR