Anti-MAPK3 Mouse Monoclonal Antibody [clone: 250603]

Supplier: R & D Systems
MAB1940
220019-474EA 600.15 USD
220019-474
Anti-MAPK3 Mouse Monoclonal Antibody [clone: 250603]
Antibodies
Type: Primary
Antigen: MAPK3
Clonality: Monoclonal
Clone: 250603
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR