Anti-FZD4 Rat Monoclonal Antibody [clone: 145901]

Supplier: R & D Systems
MAB194-050 MAB194-100
220019-476EA 616.06 USD
220019-476 220019-475
Anti-FZD4 Rat Monoclonal Antibody [clone: 145901]
Antibodies
Type: Primary
Antigen Symbol: FZD4
Antigen Name: frizzled class receptor 4
Antigen Synonyms: CD344, EVR1, FEVR, FZD4S, Fz-4, Fz4, FzE4, GPCR, hFz4
Clonality: Monoclonal
Clone: 145901
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR