Anti-PLG Mouse Monoclonal Antibody [clone: 270409]

Supplier: R & D Systems
MAB1939
220019-473EA 600.15 USD
220019-473
Anti-PLG Mouse Monoclonal Antibody [clone: 270409]
Antibodies
Type: Primary
Antigen: PLG
Clonality: Monoclonal
Clone: 270409
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR