Anti-A2M Mouse Monoclonal Antibody [clone: 257316]

Supplier: R & D Systems
MAB1938
220019-472EA 779.48 USD
220019-472
Anti-A2M Mouse Monoclonal Antibody [clone: 257316]
Antibodies
Type: Primary
Antigen: A2M
Clonality: Monoclonal
Clone: 257316
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR