Anti-MCPT7 Rat Monoclonal Antibody [clone: 263019]

Supplier: R & D Systems
MAB1937
220019-471EA 772.25 USD
220019-471
Anti-MCPT7 Rat Monoclonal Antibody [clone: 263019]
Antibodies
Type: Primary
Antigen Symbol: MCPT7
Antigen Name: Tryptase beta-1
Antigen Synonyms: MCP-7
Clonality: Monoclonal
Clone: 263019
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR