Anti-C2 Mouse Monoclonal Antibody [clone: 269716]

Supplier: R & D Systems
MAB1936
220019-470EA 594.37 USD
220019-470
Anti-C2 Mouse Monoclonal Antibody [clone: 269716]
Antibodies
Type: Primary
Antigen: C2
Clonality: Monoclonal
Clone: 269716
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR