Anti-PTPRJ Mouse Monoclonal Antibody [clone: 261922]

Supplier: R & D Systems
MAB19341
220019-468EA 561.11 USD
220019-468
Anti-PTPRJ Mouse Monoclonal Antibody [clone: 261922]
Antibodies
Type: Primary
Antigen: PTPRJ
Clonality: Monoclonal
Clone: 261922
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR