Anti-PTPRJ Mouse Monoclonal Antibody [clone: 143-41]

Supplier: R & D Systems
MAB1934
220019-467EA 633.42 USD
220019-467
Anti-PTPRJ Mouse Monoclonal Antibody [clone: 143-41]
Antibodies
Type: Primary
Antigen: PTPRJ
Clonality: Monoclonal
Clone: 143-41
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR