Anti-PTPN2 Mouse Monoclonal Antibody [clone: 252294]

Supplier: R & D Systems
MAB1930
220019-466EA 639.2 USD
220019-466
Anti-PTPN2 Mouse Monoclonal Antibody [clone: 252294]
Antibodies
Type: Primary
Antigen: PTPN2
Clonality: Monoclonal
Clone: 252294
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR