Anti-ABCC1 Mouse Monoclonal Antibody [clone: 46451]

Supplier: R & D Systems
MAB1929
220019-463EA 566.89 USD
220019-463
Anti-ABCC1 Mouse Monoclonal Antibody [clone: 46451]
Antibodies
Type: Primary
Antigen: ABCC1
Clonality: Monoclonal
Clone: 46451
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR