Anti-TMPRSS5 Rat Monoclonal Antibody [clone: 282324]

Supplier: R & D Systems
MAB1928
220019-462EA 584.25 USD
220019-462
Anti-TMPRSS5 Rat Monoclonal Antibody [clone: 282324]
Antibodies
Type: Primary
Antigen: TMPRSS5
Clonality: Monoclonal
Clone: 282324
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR