Anti-RAD17 Mouse Monoclonal Antibody [clone: 223802]

Supplier: R & D Systems
MAB1926
220019-460EA 561.11 USD
220019-460
Anti-RAD17 Mouse Monoclonal Antibody [clone: 223802]
Antibodies
Type: Primary
Antigen: RAD17
Clonality: Monoclonal
Clone: 223802
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR