Anti-SOX17 Mouse Monoclonal Antibody [clone: 245013]

Supplier: R & D Systems
MAB1924
220019-457EA 639.2 USD
220019-457
Anti-SOX17 Mouse Monoclonal Antibody [clone: 245013]
Antibodies
Type: Primary
Antigen: SOX17
Clonality: Monoclonal
Clone: 245013
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR