Anti-FRZB Mouse Monoclonal Antibody [clone: 133931]

Supplier: R & D Systems
MAB1921
220019-456EA 772.25 USD
220019-456
Anti-FRZB Mouse Monoclonal Antibody [clone: 133931]
Antibodies
Type: Primary
Antigen Symbol: FRZB
Antigen Name: frizzled-related protein
Antigen Synonyms: FRE, FRITZ, FRP-3, FRZB-1, FRZB-PEN, FRZB1, FZRB, OS1, SFRP3, SRFP3, hFIZ
Clonality: Monoclonal
Clone: 133931
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR