Anti-SFTPD Mouse Monoclonal Antibody [clone: 292201]

Supplier: R & D Systems
MAB1920
220019-454EA 600.15 USD
220019-454
Anti-SFTPD Mouse Monoclonal Antibody [clone: 292201]
Antibodies
Type: Primary
Antigen: SFTPD
Clonality: Monoclonal
Clone: 292201
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR