Anti-TMEFF1 Mouse Monoclonal Antibody [clone: 249511]

Supplier: R & D Systems
MAB1919
220019-453EA 763.57 USD
220019-453
Anti-TMEFF1 Mouse Monoclonal Antibody [clone: 249511]
Antibodies
Type: Primary
Antigen: TMEFF1
Clonality: Monoclonal
Clone: 249511
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR