Anti-FN1 Mouse Monoclonal Antibody [clone: P1F11]

Supplier: R & D Systems
MAB19181
220019-452EA 594.37 USD
220019-452
Anti-FN1 Mouse Monoclonal Antibody [clone: P1F11]
Antibodies
Type: Primary
Antigen Symbol: FN1
Antigen Name: fibronectin 1
Antigen Synonyms: CIG, ED-B, FINC, FN, FNZ, GFND, GFND2, LETS, MSF
Clonality: Monoclonal
Clone: P1F11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR