Anti-MFRP Rat Monoclonal Antibody [clone: 291218]

Supplier: R & D Systems
MAB1915
220019-447EA 561.11 USD
220019-447
Anti-MFRP Rat Monoclonal Antibody [clone: 291218]
Antibodies
Type: Primary
Antigen: MFRP
Clonality: Monoclonal
Clone: 291218
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR