Anti-ECE3 Mouse Monoclonal Antibody [clone: 195031]

Supplier: R & D Systems
MAB1914
220019-446EA 763.57 USD
220019-446
Anti-ECE3 Mouse Monoclonal Antibody [clone: 195031]
Antibodies
Type: Primary
Antigen: ECE3
Clonality: Monoclonal
Clone: 195031
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR