Anti-FGF13 (1B) Mouse Monoclonal Antibody [clone: 298923]

Supplier: R & D Systems
MAB1909
220019-445EA 561.11 USD
220019-445
Anti-FGF13 (1B) Mouse Monoclonal Antibody [clone: 298923]
Antibodies
Type: Primary
Antigen Symbol: FGF13 (1B)
Antigen Name: fibroblast growth factor 13
Antigen Synonyms: FGF-13, FGF2, FHF-2, FHF2
Clonality: Monoclonal
Clone: 298923
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR