Anti-SLAMF6 Mouse Monoclonal Antibody [clone: 292811]

Supplier: R & D Systems
MAB19081
220019-444EA 655.11 USD
220019-444
Anti-SLAMF6 Mouse Monoclonal Antibody [clone: 292811]
Antibodies
Type: Primary
Antigen: SLAMF6
Clonality: Monoclonal
Clone: 292811
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR